Learn More
Perindoprilat Isopropyl Ester tert-Butylamine Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (2S,3aS,7aS)-1-[(2S)-2-[((1S)-1-[(1Methylethoxy)carbonyl]butyl)amino]propanoyl]octahydro-1H-indole-2-carboxylic Acid tert-Butylamine Salt |
| CAS | 2771193-11-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H34 N2 O5 . C4 H11 N |
| Quantity | 50 mg |
| Synonym | (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-[(1Methylethoxy)carbonyl]butyl]amino]propanoyl]octahydro-1H-indole-2-carboxylic Acid tert-Butylamine Salt, 1H-Indole-2-carboxylic acid','octahydro-1-[(2S)-2-[[(1S)-1-[(1-methylethoxy)carbonyl]butyl]amino]-1-oxopropyl]-','(2S,3aS,7aS)-','compd. with 2-methyl-2-propanamine (1:1) (ACI), Perindopril Related Compound E (USP) |
| InChI Formula | InChI=1S/C20H34N2O5.C4H11N/c1-5-8-15(20(26)27-12(2)3)21-13(4)18(23)22-16-10-7-6-9-14(16)11-17(22)19(24)25;1-4(2,3)5/h12-17,21H,5-11H2,1-4H3,(H,24,25);5H2,1-3H3/t13-,14-,15-,16-,17-;/m0./s1 |
| SMILES | CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O)C(=O)OC(C)C.CC(C)(C)N |
| IUPAC Name | (2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-oxo-1-propan-2-yloxypentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid;2-methylpropan-2-amine |
| Molecular Weight (g/mol) | 455.63 |
| Show More |