Learn More
Perillartine, MedChemExpress
Product Code. 16479176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16479176 Supplier Medchem Express Supplier No. HYN2084
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Perillartine is a sweetener, which activates the taste receptor type 1 member 2 (Tas1r2) subunit in a species-dependent manner.
Perillartine is a sweetener, which activates the taste receptor type 1 member 2 (Tas1r2) subunit in a species-dependent manner.
Specifications
Chemical Name or Material Perillartine
CAS 30950-27-7
Color White
Purity Grade Notes Research
Molecular Formula C10H15NO
Quantity 100 mg
Synonym DL-Perillartine
Solubility Information DMSO : ≥ 100 mg/mL (605.22 mM) ∣H2O : 0.67 mg/mL (4.05 mM; Need ultrasonic)
SMILES CC(C1CC=C(/C=N/O)CC1)=C
Molecular Weight (g/mol) 165.23
Formula Weight 165.23
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism