Learn More
Invitrogen™ Pericentrin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554109
Description
Immunogen sequence: MLDLSSWSSP EVLRKDWTLE PWPSLPVTPH SGALSLCSAD TSLGDRADTS LPQTQGPGLL CSPGVSAAAL ALQWAESPPA DDHHVQRTAV EKDVEDFITT SFDSQETLSS PPPGLEGKAD RSEKSDGSG Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 58%.
This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.
Specifications
| Pericentrin | |
| Polyclonal | |
| Unconjugated | |
| PCNT | |
| AW476095; C86676; KEN; kendrin; KIAA0402; m239Asp; m275Asp; MOPD2; PCN; Pcnt; PCNT2; PCNTB; PCTN2; Pericentrin; pericentrin (kendrin); pericentrin 2; pericentrin-2; pericentrin-250; pericentrin-360; pericentrin-380; pericentrin-B; SCKL4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5116 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95613 | |
| PCNT | |
| Recombinant protein corresponding to Human Pericentrin. Recombinant protein control fragment (Product #RP-91791). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.