Learn More
Perfluoro-2,5-dimethyl-3,6-dioxanonanoic Acid, TRC
Product Code. 17971425
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971425 500 mg 500mg
17934255 50 mg 50mg
17901075 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17971425 Supplier Toronto Research Chemicals Supplier No. TRCP286243500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13252-14-7
Molecular Formula C9 H F17 O4
Molecular Weight (g/mol) 496.0747
Synonym Propanoic acid, 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(heptafluoropropoxy)propoxy]- (9CI), Propionic acid, 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(heptafluoropropoxy)propoxy]- (7CI,8CI), 2,3,3,3-Tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoic acid, 2,3,3,3-Tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]-1-propanoic acid, X 70-540-3, Perfluoro-2,5-dimethyl-3,6-dioxanonanoic Acid (HFPO-TA)
IUPAC Name 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoic acid
SMILES OC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C9HF17O4/c10-2(1(27)28,5(14,15)16)29-9(25,26)4(13,7(20,21)22)30-8(23,24)3(11,12)6(17,18)19/h(H,27,28)
Formula Weight 495.9603
Percent Purity >95%
Chemical Name or Material Perfluoro-2,5-dimethyl-3,6-dioxanonanoic Acid (HFPO-TA)
Grade HPLC