missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pepsin From Porcine Stomach Mucosa

Product Code. 11468220 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
This item is not returnable. View return policy

Product Code. 11468220

Brand: MP Biomedicals™ 0210259850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond

Pepsin contains the “cathepsin” component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity.

  • CAS Number: 9001-75-6
  • EC Number: 232-629-3
  • MDL Number: MFCD00081840
  • Format: off white lyophilized powder
  • Specific activity: ≥1:10000

Used in preparation of Fab fragments from antibodies and to hydrolyze dry cervical samples in mice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Molecular Biology, Proteomics
Molecular Weight (g/mol) ∽35000
Name Pepsin
pH Range 2.5 to 4.5
Quantity 50 g
Source Porcine
Storage Requirements 15° to 30°C
Form Powder
compliance-icons compliance-icons
Product Identifier
  • Pepsin, 1:10,000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : pepsin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.