Learn More
Invitrogen™ PEO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592383
Description
Immunogen sequence: GAREGFLLSK APEFEDSEEV RRIWNRAIPL WELPDQEEVQ LADTMFGLTK VTDDTLKRFS VRYLRPARSL VFPWFSPGGS GLRGLKLLEA KCQGDGVSYE ETTIPRPSAY HNLFGLPLIS RRDAEVVLTS RELDSLALNQ STGLPTLTLP RGTTCLPPAL LPYLEQFRRI VFWLGDDLRS WEAAKLFARK LNPKRCFLVR P Positive Samples: MCF7; Cellular Location: Mitochondrion matrix, mitochondrion nucleoid.
Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars.Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars (Spelbrink et al., 2001 [PubMed 11431692]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
| PEO1 | |
| Polyclonal | |
| Unconjugated | |
| TWNK | |
| ataxin 8; Ataxin8; ATXN 8; ATXN8; C10 orf2; C10orf 2; C10orf2; C18H10orf2; C1H10orf2; C26H10orf2; C28H10orf2; chromosome 10 open reading frame 2; D19Ertd626e; fi04d09; FLJ21832; IOSCA; mitochondrial twinkle protein; MTDPS7; PEO; PEO 1; peo1; PEOA3; PRLTS5; progressive external ophthalmoplegia 1; progressive external ophthalmoplegia 1 (human); progressive external ophthalmoplegia 1 homolog; progressive external ophthalmoplegia 1 protein; Progressive external ophthalmoplegia 1 protein homolog; SANDO; SCA 8; SCA8; T7 gp4-like protein with intramitochondrial nucleoid localization; T7 helicase-related protein with intramitochondrial nucleoid localization; T7-like mitochondrial DNA helicase; twinkle; Twinkle mtDNA helicase; twinkle protein, mitochondrial; twinkle protein, mitochondrial-like; Twinkle protein, mitochondrial-like protein; Twinl; Twnk; wu:fi04d09 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 226153, 309441, 56652 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.92 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q8CIW5, Q96RR1 | |
| TWNK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 135-335 of human C10orf2 (NP_0686022). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.