Learn More
Pentafluorophenyl trifluoroacetate, 98%
Product Code. p-4228862 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. 10526640
Brand: Thermo Scientific Acros 431520050
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 14533-84-7 | C8F8O2 | 280.07 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14533-84-7
Molecular Formula C8F8O2
Molecular Weight (g/mol) 280.07
MDL Number MFCD00134438
InChI Key VCQURUZYYSOUHP-UHFFFAOYSA-N
Synonym pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8
PubChem CID 4327891
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
SMILES FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F
Specifications
Melting Point 3.0°C
Color Colorless
Boiling Point 122.0°C to 125.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 280.08
Percent Purity 98%
Physical Form Oil
Chemical Name or Material Pentafluorophenyl trifluoroacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentafluorphenyl trifluoracetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.