Learn More
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate, 97%, Thermo Scientific™
Product Code. p-3541037
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11356815 250 mg Amber Glass Bottle 250mg
10738075 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11356815 Supplier Thermo Scientific Maybridge Supplier No. CC58326CB
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Chemical Identifiers
CAS 910036-96-3
Molecular Formula C17H12F5NO4
Molecular Weight (g/mol) 389.28
MDL Number MFCD09702389
InChI Key DOADFHHFWSVLLM-UHFFFAOYSA-N
Synonym pentafluorophenyl 6-tetrahydropyran-4-yloxy nicotinate, 2,3,4,5,6-pentafluorophenyl 6-oxan-4-yloxy pyridine-3-carboxylate, pentafluorophenyl 6-oxan-4-yloxy pyridine-3-carboxylate, pentafluorophenyl 6-oxan-4-yl oxy pyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-oxan-4-yloxy pyridine-3-carboxylate, 6-4-oxanyloxy-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229582
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 6-(oxan-4-yloxy)pyridine-3-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CN=C(OC3CCOCC3)C=C2)C(F)=C1F
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 389.27
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate