Learn More
Pentaerythritol-2-13C, CDN
Product Code. 30318641
Change view
Click to view available options
Quantity:
0.1 g
Unit Size:
0.1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318641 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30318641 Supplier CDN Supplier No. CDNC32790.1G
Please to purchase this item. Need a web account? Register with us today!
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pentaerythritol-2-13 C
CAS 334974-06-0
Molecular Formula 13 C C4 H12 O4
Quantity 0.1 g
Synonym 1,3-Propanediol-2-13 C, 2,2-bis(hydroxymethyl)- (9 CI), Pentaerythritol-2-13 C, 2,2-Bis(hydroxymethyl)-1,3-propanediol-2-13 C, Tetra(hydroxymethyl)methane-13 C, Tetrakis(hydroxymethyl)methane-13 C, Pentaerythritol-13 C (methane-13 C)
InChI Formula InChI=1 S/C5H12O4/c6-1-5(2-7,3-8)4-9/h6-9 H,1-4H2/i5+1
SMILES OC[13 C](CO)(CO)CO
IUPAC Name 2,2-bis(hydroxymethyl)(2 ^{13}C)propane-1,3-diol
Molecular Weight (g/mol) 137.139
Formula Weight 137.077 g/mol
Percent Purity 99 atom % 13 C, min 98% Chemical Purity