missing translation for 'onlineSavingsMsg'
Learn More

Penicillin V Potassium Salt, TRC

Product Code. 17922895
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922895 100 mg 100mg
30132718 25 g 25g
17983325 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17922895 Supplier Toronto Research Chemicals Supplier No. TRCP223500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-98-9
Molecular Formula C16 H17 N2 O5 S . K
Molecular Weight (g/mol) 388.48
Synonym Phenoxymethylpenicillin Potassium, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid potassium salt (1:1), 3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, [2S-(2α,5α,6β)]-3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, Abbocillin VK, Acipen, Anapenil, Antibiocin, Apo-Pen VK, Apsin VK, Arcacil, Arcasin, Asillin, Beapen, Beepen Vk, Beromycin, Beromycin (penicillin), Beromycin 400, Betapen V, Betapen Vk, Calciopen K, Cilacil, Cilicaine VK, Cliacil, Compocillin VK, DQV-K, Distakaps V-K, Distaquaine V-K, Dowpen V-K, DuraPenicillin, Fenocin, Fenocin Forte, Fenospen, Fenoxcillin, Fenoxypen, Icipen, Isocillin, Ispenoral, Kavepenin, L.P.V., Ledercillin Vk, Len VK, Megacilina Oral, Megacillin-oral, Microcillin VK, Micropen VK, Milcopen, Monocillin E, Nadopen V, Newcillin, Novo VK, Novopen VK, Ocillin VK, Oracil VK, Oracillin VK, Orapen, Orvek, Ospen 250, Ospen KV, Ospeneff, P.V.O., PVK, Pedipen, Pen V-K, Pen-V, Pen-Vee K Powder, Pen-Vee-K, Pen-Vi-K, Penadur VK Mega, Penagen, Penapar VK, Pencompren, Penicillin V potassium, Penicillin V potassium salt, Penicillin VK, Penivoral, Penoxilin, Pentabs, Pentacillin, Pentranex, Penvikal, Penvisil, Pfizerpen Vk, Phenoxymethylpenicillin potassium, Phenoxymethylpenicillin potassium salt, Potassium 2,2-dimethyl-6α-phenoxyacetamidopenam-3α-carboxylate, Potassium penicillin V, Potassium penicillin V salt, Potassium phenoxymethylpenicillin, Potassium phenoxymethylpenicillinate, Primcillin, Qidpen VK, Rafapen V-K, Robicillin VK, Rocilin, Rocillin VK, Roscopenin, SK-Penicillin VK, Servipen V, Stabilin V-K, Stabillin VK syrup 125, Stabillin VK syrup 62.5, Sumapen VK, Suspen, Trepopen, Trepopen VK, Uticillin Vk, V-Cil K, V-Cillin K, V-Kal K, V-Pen, V-Penicillin Kalium, Vamosyn, Veetids, Vepen, Vepicombin
IUPAC Name potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COc3ccccc3)C(=O)N2[C@H]1C(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H18N2O5S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)24-16)17-10(19)8-23-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1
Formula Weight 388.0495
Percent Purity >95%
Chemical Name or Material Phenoxymethylpenicillin Potassium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.