Learn More
UVP Pen-Ray™ Gas Lamp Filll
Click to view available options
Gas Fill:
Neon
Neon-mercury
Xenon
Unit Size:
Each
Specifications
Specifications
| Gas Fill | Neon |
| Voltage | 450 V |
| Wavelength | 254 nm |
| Length (Metric) | 117.3 mm |
| Material | Quartz |
| Product Type | Pen Ray |