missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pemirolast potassium, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.
Specifications
Specifications
| Chemical Name or Material | Pemirolast potassium |
| Physical Form | Solid |
| CAS | 100299-08-9 |
| Color | Yellowish Green |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H7KN6O |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | TWT-8152 BMY 26517 |
| Solubility Information | H2O : 50 mg/mL (187.76 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction