Learn More
Pemirolast potassium, MedChemExpress
Product Code. 16478518
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16478518
Brand: Medchem Express HYB0538A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.
Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.
Specifications
Chemical Name or Material Pemirolast potassium
Physical Form Solid
CAS 100299-08-9
Color Yellowish Green
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H7KN6O
For Use With (Application) COVID-19-immunoregulation
Synonym TWT-8152 BMY 26517
Solubility Information H2O : 50 mg/mL (187.76 mM; Need ultrasonic)
SMILES O=C1C(C2=N[N-]N=N2)=CN=C3N1C=CC=C3C.[K+]
Molecular Weight (g/mol) 266.3
Formula Weight 266.3
Percent Purity 99.93%
Grade Research