missing translation for 'onlineSavingsMsg'
Learn More

Pemetrexed disodium, MedChemExpress

Product Code. 16421006
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421006 200 mg 200mg
16485136 100 mg 100mg
16470527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16421006 Supplier Medchem Express Supplier No. HY10820A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pemetrexed disodium (LY231514 disodium) is an antifolate, the Kis of the pentaglutamate of Pemetrexed disodium are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.

Pemetrexed disodium (LY231514 disodium) is an antifolate, the Kis of the pentaglutamate of Pemetrexed disodium are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.

Specifications

Chemical Name or Material Pemetrexed disodium
CAS 150399-23-8
Color White
Purity Grade Notes Research
Molecular Formula C20H19N5Na2O6
Quantity 200 mg
Synonym LY231514 disodium
Solubility Information H2O : ≥ 100 mg/mL (212.15 mM)
SMILES O=C1NC(N)=NC2=C1C(CCC3=CC=C(C=C3)C(N[C@@H](CCC(O[Na])=O)C(O[Na])=O)=O)=CN2
Molecular Weight (g/mol) 471.37
Formula Weight 471.37
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.