missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PEG3 Monoclonal antibody specifically detects PEG3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | PEG3 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI1E10 |
| Conjugate | PE |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | PBS |
| Gene Alias | DKFZp781A095, KIAA0287PW1, paternally expressed 3, paternally expressed gene 3, Zinc finger and SCAN domain-containing protein 24, ZNF904, ZSCAN24Kruppel-type zinc finger protein |
| Host Species | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 1-364 of human PEG3 (NP_006201) produced in E.coli. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?