Learn More
Pefloxacin-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Pefloxacin-d3 |
| CAS | 2733455-58-6 |
| Molecular Formula | C17 D3 H17 F N3 O3 |
| Quantity | 10 mg |
| Synonym | 1-Ethyl-6-fluoro-1,4-dihydro-7-[4-(methyl-d3)-1-piperazinyl]-4-oxo-3-quinolinecarboxylic acid, Pefloxacin-D3 (N-methyl-D3), Pefloxacin-d3 |
| InChI Formula | InChI=1S/C17H20FN3O3/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21/h8-10H,3-7H2,1-2H3,(H,23,24)/i2D3 |
| SMILES | [2H]C([2H])([2H])N1CCN(CC1)c2cc3N(CC)C=C(C(=O)O)C(=O)c3cc2F |
| IUPAC Name | 1-ethyl-6-fluoro-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 336.376 |
| Formula Weight | 336.168 |