missing translation for 'onlineSavingsMsg'
Learn More

PDLIM7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen PDLIM7
Concentration 0.15 mg/mL
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16876914
View Documents
Proteintech
17080-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16866914
View Documents
Proteintech
17080-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants.
Specifications

Specifications

PDLIM7
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Rat, Human
Q3TJD7, Q9NR12, Q9Z1Z9
286908, 67399, 9260
PDLIM7 Fusion Protein Ag1334
Primary
-20°C
PDLIM7
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ENIGMA, LIM mineralization protein, LMP, LMP1, PDLIM7, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7, Protein enigma
Pdlim7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.