missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (MM0004-8A89) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60969
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PDGFR alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100-400, Immunohistochemistry-Paraffin 1:100-1:400 | |
| P16234 | |
| PDGFRA | |
| Human recombinant PDGFR alpha (N-terminal) | |
| 0.1 mg | |
| Protein Kinase | |
| 5156 | |
| Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0004-8A89 | |
| Unconjugated | |
| Lyophilized from a 0.2 um filtered solution in PBS, 0.025 mg size is provided in liquid form. with No Preservative | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| No cross-reactivity is shown with human PDGF Receptor beta | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu