missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576570
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Removes the formyl group from the N-terminal Met of newly synthesized proteins.
Specifications
| Polyclonal | |
| Unconjugated | |
| 2610019N19Rik; PDF; PDF1A; peptide deformylase (mitochondrial); peptide deformylase, mitochondrial; peptide deformylase-like protein; peptide deformylase-like protein-like; polypeptide deformylase; similar to peptide deformylase-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64146, 68023, 690214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBH1 | |
| Recombinant protein of human PDF. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction