Learn More
PD318088, MedChemExpress
Product Code. 16495736
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16495736
Brand: Medchem Express HY12062
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PD318088 is a potent, allosteric and non-ATP competitive MEK1/2 inhibitor, an analog of PD184352 (HY-50295). PD318088 binds simultaneously with ATP in a region of the MEK1 active site that is adjacent to the ATP-binding site. PD318088 can be used for cancer research.
PD318088 is a potent, allosteric and non-ATP competitive MEK1/2 inhibitor, an analog of PD184352 (HY-50295). PD318088 binds simultaneously with ATP in a region of the MEK1 active site that is adjacent to the ATP-binding site. PD318088 can be used for cancer research.
Specifications
Chemical Name or Material PD318088
CAS 391210-00-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H13BrF3IN2O4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (178.22 mM)
SMILES BrC1=CC(C(NOCC(CO)O)=O)=C(C(F)=C1F)NC2=CC=C(C=C2F)I
Molecular Weight (g/mol) 561.09
Formula Weight 561.09
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease