missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PD 407824
GREENER_CHOICE

Product Code. 18055262
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 18055262

Brand: Tocris Bioscience™ 2694/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of Chk1 and Wee1

Selective inhibitor of checkpoint kinases Chk1 and Wee1 (IC50 values are 47 and 97 nM respectively). Displays selectivity over a range of other protein kinases; IC50 values are 3.4, 3.75, > 5, > 50, > 50 and > 50 μM for PKC, CDK4, other CDKs, c-Src, PDGFR and FGFR respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622864-54-4
Molecular Formula C20H12N2O3

Specifications

Quantity 10 mg
Formula Weight 328.32
Purity >98%
Molecular Weight (g/mol) 328.33
Chemical Name or Material PD 407824
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.