Learn More
PD-146176, TRC
Specifications
Specifications
| Chemical Name or Material | PD-146176 |
| CAS | 4079-26-9 |
| Molecular Formula | C15H11NS |
| Quantity | 25 mg |
| Synonym | 6,11-Dihydro[1]benzothiopyrano[4,3-b]indole, NSC 168807, PD 146176 |
| InChI Formula | InChI=1S/C15H11NS/c1-3-7-13-10(5-1)12-9-17-14-8-4-2-6-11(14)15(12)16-13/h1-8,16H,9H2 |
| SMILES | C1(N2)=C(CSC3=CC=CC=C31)C4=C2C=CC=C4 |
| IUPAC Name | 6,11-dihydrothiochromeno[4,3-b]indole |
| Molecular Weight (g/mol) | 237.32 |
| Formula Weight | 237.0612 |