Learn More
Greiner Bio-One PCR Tube, Thin Wall, Attached Domed Cap
Description
- Manufactured out of medical-grade polypropylene without interfering additives
- Attached, domed cap
- Color-coded
- Thin-walled for efficient heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Suitable for real-time / quantitative PCR
- Comprehensive thermal cycler compatibility
Specifications
Specifications
| Color | Yellow |
| Volume (Metric) | 500 μL |
| Material | Polypropylene |
| Dnase Free | Yes |
| RNase Free | Yes |
| Closure Included | Polypropylene |
| Product Type | Thin Wall |