Learn More
Greiner Bio-One PCR Tube, Thin Wall, Attached Domed Cap
Product Code. 15995321
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
15995321 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15995321 Supplier Greiner Bio-One Supplier No. 672276
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Free of detectable DNase, RNase, human DNA, endotoxin, and PCR inhibitors.
  • Manufactured out of medical-grade polypropylene without interfering additives
  • Attached, domed cap
  • Color-coded
  • Thin-walled for efficient heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Suitable for real-time / quantitative PCR
  • Comprehensive thermal cycler compatibility
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Volume (Metric) 500 μL
Material Polypropylene
Dnase Free Yes
RNase Free Yes
Closure Included Polypropylene
Product Type Thin Wall