missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One PCR Strip Tubes without Cap

Product Code. 10013391
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Unit Size:
Pack of 125
Pack of 1250
This item is not returnable. View return policy

Product Code. 10013391

Brand: Greiner Bio-One 673277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in research. Greiner Bio-One PCR Strip Tubes without Cap are free of detectable DNase, Rnase and human DNA.

  • Non-Pyrogenic
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
TRUSTED_SUSTAINABILITY

Specifications

Color Violet
Volume (Metric) 0.2 mL
For Use With (Equipment) PCR
Sterility Non-sterile
Material Polypropylene
Product Type PCR Tube Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.