Learn More
R&D Systems™ pCPA methyl ester hydrochloride
Description
pCPA methyl ester hydrochloride is a tryptophan hydroxylase inhibitor. Inhibits 5-HT synthesis. Depletes 5-HT levels in orbitofrontal cortex and impairs reversal learning in rats. Methyl ester of p-Chlorophenylalanine (Cat. No. 0938).
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-DL-Phenylalanine methyl ester hydrochloride |
| CAS | 14173-40-1 |
| Quantity | 100 mg |
| Product Line | Tocris |
| Target | Hydroxylase Inhibitors |
| Molecular Formula | C10H12ClNO2.HCl |
| Formula Weight | 250.12 |
| Purity | >98% |