Learn More
Tocris Bioscience™ pCPA methyl ester hydrochloride
Tryptophan hydroxylase inhibitor
Brand: Tocris Bioscience™ 5871/100
This item is not returnable.
View return policy
Description
pCPA methyl ester hydrochloride is a tryptophan hydroxylase inhibitor. Inhibits 5-HT synthesis. Depletes 5-HT levels in orbitofrontal cortex and impairs reversal learning in rats. Methyl ester of p-Chlorophenylalanine (Cat. No. 0938).Specifications
| 4-Chloro-DL-Phenylalanine methyl ester hydrochloride | |
| 14173-40-1 | |
| 100 mg |