Learn More
Tocris Bioscience™ PCI 29732
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PCI 29732 is a potent BTK inhibitor (IC50 = 0.3 nM). Blocks B cell antigen receptor (BCR)-mediated gene expression in CD20+ B cells.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | PCI 29732 |
| Purity | >99% |
| Molecular Weight (g/mol) | 371.44 |
| Inhibitors | PCI 29732 |