missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCDHB15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17770784
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17770784 200 μL 200µL
17793094 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17770784 Supplier Invitrogen™ Supplier No. PA5119381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCDHB15 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHB15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PCDHB15
Gene Accession No. Q9Y5E8
Gene Alias PCDHB15; PCDH-BETA15; PCDH-beta-15; protocadherin beta 15; protocadherin beta-15
Gene Symbols PCDHB15
Host Species Rabbit
Immunogen Recombinant protein Protocadherin Beta 15. The antigen corresponds to amino acid range 35-347 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56121, 93886
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.