Learn More
Lenz Laborglasinstrumente™ PBT Red Screw Caps
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Screw thread tubes |
| Includes | DIN thread (GL) |
| Material | PBT |
| Max. Temperature (Metric) | 180°C |
| Quantity | 10Pack |
| Screw Material | PBT |
| Product Line | Cap |
| Thread Size (Metric) | GL 18 |
| Product Type | Screw Cap |