Learn More
PBK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16110-1-AP-20UL
This item is not returnable.
View return policy
Description
This genes encodes a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.Specifications
| PBK | |
| Polyclonal | |
| Unconjugated | |
| PBK | |
| Cancer/testis antigen 84, CT84, MAPKK like protein kinase, Nori 3, PBK, PDZ binding kinase, SPK, TOPK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 290326, 52033, 55872 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96KB5, Q9JJ78 | |
| Pbk | |
| PBK Fusion Protein Ag9033 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |