missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax6 Antibody (PAX6/1166), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47915AF532
This item is not returnable.
View return policy
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Intracellular), CyTOF, Gene Knock-Out
Specifications
| Pax6 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367) | |
| 0.1 mL | |
| Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 5080 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining), CyTOF | |
| PAX6/1166 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction