missing translation for 'onlineSavingsMsg'
Learn More

Pax2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

227.00 € - 326.00 €

Specifications

Antigen Pax2
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:2500-1:10000
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18631546
View Documents
Novus Biologicals
NBP2-99012-100ul
100 μg
326.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18667735
View Documents
Novus Biologicals
NBP2-99012-50ul
50 μg
227.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pax2 Polyclonal antibody specifically detects Pax2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Pax2
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers
0.2 μm filtered solution in PBS
5076
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:2500-1:10000
Polyclonal
Purified
RUO
Human, Mouse
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Pax2.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.