missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress MC1R U2OS β-Arrestin GPCR Assay

Product Code. 17333919
Encompass
Change view
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17333919 200 dp (2 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17333919 Supplier DiscoverX Supplier No. 930861E3CP5M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress MC1R U2OS β-Arrestin GPCR Assay measures MC1R (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ MC1R β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. α-MSH) based activation of MC1R GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged MC1R and EA-tagged β-Arrestin 2. Activation of MC1R stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry β-Arrestin-2 recruitment
For Use With (Application) Assay-Ready eXpress Kits
Primary Agonist Melanotan II
Quantity 200 dp (2 x 96-well)
Target Entry MC1R
Cell Line U2OS
Target MC1R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.