Learn More
Thermo Scientific™ Particle-Certified Glass Containers
Product Code. 16474390
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Product Type:
Boston Round Bottle
Jug
Packer Bottle
Sample Jar
Color:
Amber
Clear
No. per Case:
4
12
24
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 4
Product Code. 16474390
Brand: Thermo Scientific™ 114125CT/LP
Only null left
Add to basket
Add to basket
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
  • Low particle applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Clear
Description Clear Glass Boston Round Bottle with Closed-top Closure, 125 mL
Diameter (English) 1.875 in.
Material Glass
No. per Case 12
Height (Metric) 111.13 mm
Product Type Boston Round Bottle
Height (English) 4.375 in.
Closure Liner PTFE/Silicone
Closure Material Polypropylene, Low-shedding
Closure Size 24-414 mm
Diameter (Metric) 47.63 mm
Closure Type Closed-top