missing translation for 'onlineSavingsMsg'
Learn More

Parthenolide, MedChemExpress

Product Code. 16401496
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16401496

Brand: Medchem Express HYN0141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Parthenolide is a sesquiterpene lactone found in the medicinal herb Feverfew. Parthenolide exhibits anti-inflammatory activity by inhibiting NF-κB activation; also inhibits HDAC1 protein without affecting other class I/II HDACs.

Parthenolide is a sesquiterpene lactone found in the medicinal herb Feverfew. Parthenolide exhibits anti-inflammatory activity by inhibiting NF-κB activation

Specifications

Chemical Name or Material Parthenolide
CAS 20554-84-1
Color White
Purity Grade Notes Research
Molecular Formula C15H20O3
Quantity 200 mg
Synonym (-)-Parthenolide
Solubility Information DMSO : ≥ 100 mg/mL (402.71 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C/C1=C\CC[C@@](C)(O2)[C@H]2[C@@H](OC(C3=C)=O)[C@H]3CC1
Molecular Weight (g/mol) 248.32
Formula Weight 248.32
Percent Purity 97.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.