missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD6G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30757
This item is not returnable.
View return policy
Description
PARD6G Polyclonal specifically detects PARD6G in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PARD6G | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9BYG4 | |
| PARD6G | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LGSSGPPSDGTAGFVGPPAPRVLQNFHPDEAESDEDNDVVIEGTLEPARP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Par-6 (Partitioning Defective 6, C.Elegans) Homolog Gamma, Par-6 Family Cell Polarity Regulator Gamma, PAR-6 Gamma, PAR-6 Gamma Protein, Par-6 Partitioning Defective 6 Homolog Gamma, Par-6 Partitioning Defective 6 Homolog Gamma (C. Elegans), PAR6G, PAR-6G, PAR6gamma, PARD6G, Partitioning Defective 6 Homolog Gamma | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84552 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction