Learn More
Carl Roth™ Paraplast f. d. Histology
Product Code. 10566465
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
10566465 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10566465 Supplier Carl Roth™ Supplier No. X880.1
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Specifications
Packaging Type Paper Bag
Quantity 1 kg
Material Highly purified paraffin containing plastic polymers of regulated molecular weights
Form Pellet