Learn More
PAPPA Monoclonal Antibody (PAPP A 1-24)
GREENER_CHOICE
PAPPA Monoclonal Antibody (PAPP A 1-24)
Brand:  Invitrogen ABS0062402
Product Code. 11554112
  • 568.00 € / 400µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAPPA
Monoclonal
1.04 mg/mL
PBS with 15mM sodium azide; pH 7.4
Q13219
PAPPA
Purified human PAPP-A.
400 μL
Primary
Human
Antibody
IgG1 κ
ELISA, Immunohistochemistry (Paraffin), Western Blot
PAPP A 1-24
Unconjugated
PAPPA
8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1
Mouse
Protein A
RUO
5069
4°C, store in dark