Learn More
Panaxatriol, TRC
Specifications
Specifications
| Chemical Name or Material | Panaxatriol |
| CAS | 32791-84-7 |
| Molecular Formula | C30 H52 O4 |
| Quantity | 10 mg |
| Synonym | Dammarane-3,6,12-triol','20,25-epoxy-','(3β,6β,12β,20R)-, Dammarane-3β,6β,12β-triol','20,25-epoxy-','(20R)- (8CI), Panaxatriol (7CI), (3β,6β,12β,20R)-20,25-Epoxydammarane-3,6,12-triol, NSC 308880 |
| InChI Formula | InChI=1S/C30H52O4/c1-25(2)12-9-13-30(8,34-25)18-10-15-28(6)23(18)19(31)16-21-27(5)14-11-22(33)26(3,4)24(27)20(32)17-29(21,28)7/h18-24,31-33H,9-17H2,1-8H3/t18-,19+,20+,21+,22-,23-,24-,27+,28+,29+,30+/m0/s1 |
| SMILES | CC1(C)CCC[C@@](C)(O1)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5[C@H](O)C[C@@]34C |
| IUPAC Name | (3S,5R,6R,8R,9R,10R,12R,13R,14R,17S)-4,4,8,10,14-pentamethyl-17-[(2R)-2,6,6-trimethyloxan-2-yl]-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-3,6,12-triol |
| Molecular Weight (g/mol) | 476.73 |
| Formula Weight | 476.3866 |