Learn More
Invitrogen™ Pan SHANK Monoclonal Antibody (N23b/49), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545439
Description
1 μg/mL of MA5-45439 was sufficient for detection of Shank1-4 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 160kDa. Recognizes Shank1, 2 and 3.
Shank proteins make up a family of scaffold proteins identified through their interaction with a variety of membrane and cytoplasmic proteins. Shank proteins at postsynaptic sites of excitatory synapses play roles in signal transmission into the postsynaptic neuron. Studies suggest that Shank2 is expressed in the neurons of the developing retina, and could play a role in the neuronal differentiation of the developing retina. Other recent studies suggest that the disruption of glutamate receptors at the Shank postsynaptic platform could contribute to the destruction of the postsynaptic density, which underlies the synaptic dysfunction and loss in Alzheimer's disease.
Specifications
| Pan SHANK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| D3YZU1, Q4ACU6, Q80Z38, Q9BYB0, Q9JLU4, Q9QX74, Q9UPX8, Q9WV48, Q9Y566 | |
| Shank1, SHANK2, Shank3 | |
| Fusion protein amino acids 84-309 of rat Shank2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, Microarray | |
| N23b/49 | |
| FITC | |
| Shank3 | |
| AI841104; AUTS17; cortactin SH3 domain-binding protein; cortactin-binding protein 1; CORTBP1; CTTNBP1; DEL22q13.3; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; KIAA1022; Kiaa1650; mKIAA1022; proline rich synapse associated protein 1; proline rich synapse associated protein 2; proline-rich synapse associated protein 1; proline-rich synapse-associated protein 1; Proline-rich synapse-associated protein 2; ProSAP1; Prosap2; PSAP2; SAPAP-interacting protein synamon; SCZD15; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains 2; SH3 and multiple ankyrin repeat domains 3; SH3 and multiple ankyrin repeat domains protein 1; SH3 and multiple ankyrin repeat domains protein 2; SH3 and multiple ankyrin repeat domains protein 3; SH3 and multiple ankyrin repeat domains protein 3 variant c-3; SH3 and multiple ankyrin repeat domains protein 3 variant c-4; SH3 and multiple ankyrin repeat domains-like protein 3 variant 1; SH3 and multiple ankyrin repeat domains-like protein 3 variant 2; SH3 and multiple ankyrin repeat domains-like protein 3 variant 3; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-3; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-2; SH3/ankyrin domain gene 1; SH3/ankyrin domain gene 2; SH3/ankyrin domain gene 3; SHANK; shank postsynaptic density protein; Shank postsynaptic density protein 3a; SHANK1; Shank1a; Shank2; Shank3; shank3 splice variant II; shank3 splice variant III; shank3 splice variant IV; Shank3b; SHANK3c-3; SHANK3c-4; shank3e splice variant III; shank3e splice variant IV; somatostatin receptor-interacting protein; Spank1; SPANK-1; SPANK-2; SPANK-3; SSTR-interacting protein; Sstrip; synamon | |
| Mouse | |
| Protein G | |
| RUO | |
| 171093, 210274, 22941, 243961, 50944, 58234, 59312, 78957, 85358 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.