missing translation for 'onlineSavingsMsg'
Learn More

Pamiparib, MedChemExpress

Product Code. 16434966
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434966 100 mg 100mg
16493938 10 mg 10mg
16476657 5 mg 5mg
16460297 50 mg 50mg
16402926 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16434966 Supplier Medchem Express Supplier No. HY104044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pamiparib (BGB-290) is an orally active, potent, highly selective PARP inhibitor, with IC50 values of 0.9 nM and 0.5 nM for PARP1 and PARP2, respectively. Pamiparib has potent PARP trapping, and capability to penetrate the brain, and can be used for the research of various cancers including the solid tumor.

Pamiparib (BGB-290) is an orally active, potent, highly selective PARP inhibitor, with IC50 values of 0.9 nM and 0.5 nM for PARP1 and PARP2, respectively. Pamiparib has potent PARP trapping, and capability to penetrate the brain, and can be used for the research of various cancers including the solid tumor.

Specifications

Chemical Name or Material Pamiparib
CAS 1446261-44-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H15FN4O
Quantity 100 mg
Synonym BGB-290
Solubility Information DMSO : 62.5 mg/mL (209.51 mM; Need ultrasonic)
SMILES O=C1NN=C2CN(CCC3)[C@@]3(C)C(N4)=C2C5=C4C=C(F)C=C51
Molecular Weight (g/mol) 298.31
Formula Weight 298.31
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.