Learn More
Pamicogrel, MedChemExpress
Product Code. p-200097811
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16482418
Brand: Medchem Express HYU00175
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor.
Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor.
Specifications
Chemical Name or Material Pamicogrel
CAS 101001-34-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H24N2O4S
Quantity 5 mg
Synonym KBT3022
Solubility Information DMSO : 83.33 mg/mL (185.78 mM; Need ultrasonic)
SMILES O=C(OCC)CN1C(C2=NC(C3=CC=C(OC)C=C3)=C(C4=CC=C(OC)C=C4)S2)=CC=C1
Molecular Weight (g/mol) 448.53
Formula Weight 448.53
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation