Learn More
Paliperidone-d4 (Major), TRC
Spezifikation
Spezifikation
| Chemical Name or Material | rac 9-Hydroxy Risperidone-d4 |
| CAS | 1020719-55-4 |
| Molecular Formula | C23 2H4 H23 F N4 O3 |
| Quantity | 1 mg |
| Synonym | Paliperidone-(ethyl-D4), 9-Hydroxyrisperidone-(ethyl-D4) |
| InChI Formula | InChI=1S/C23H27FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h4-5,13,15,19,29H,2-3,6-12H2,1H3/i8D2,12D2 |
| SMILES | [2H]C([2H])(N1CCC(CC1)c2noc3cc(F)ccc23)C([2H])([2H])C4=C(C)N=C5C(O)CCCN5C4=O |
| IUPAC Name | 9-hydroxy-2-methyl-3-[1,1,2,2-tetradeuterio-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one |
| Molecular Weight (g/mol) | 430.51 |
| Formula Weight | 430.2318 |