Learn More
Palifosfamide, MedChemExpress
Product Code. 16482307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482307 50 mg 50mg
16418777 5 mg 5mg
16416178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16482307 Supplier Medchem Express Supplier No. HY14798
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Palifosfamide is a novel DNA alkylator and the active metabolite of ifosfamide, with antitumor activity.
Palifosfamide is a novel DNA alkylator and the active metabolite of ifosfamide, with antitumor activity.
Specifications
Chemical Name or Material Palifosfamide
Physical Form Solid
CAS 31645-39-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C4H11Cl2N2O2P
For Use With (Application) Cancer-programmed cell death
Synonym Isophosphoramide mustard IPM ZIO-201
Solubility Information DMSO : 40 mg/mL (180.98 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (64.65 mM; Need ultrasonic)
SMILES O=P(NCCCl)(NCCCl)O
Molecular Weight (g/mol) 221.02
Formula Weight 221.02
Percent Purity 95.0%
Grade Research