Learn More
Invitrogen™ Pacmilimab Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542284
Product Code. 17758903
  • 524.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Pacmilimab is a biosimilar that targets CD274.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pacmilimab
Recombinant Monoclonal
Unconjugated
CX-072
Human CD274/PD-L1/B7-H1.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
1 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A
RUO
Human
Antibody
IgG4 κ