missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel MP Biomedicals

Product Code. p-3179092 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
100 mg
Unit Size:
100mg
5mg
This item is not returnable. View return policy

Product Code. 11464862

Brand: MP Biomedicals 0219353205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 373-49-9
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00004437
InChI Key SECPZKHBENQXJG-FPLPWBNLSA-N
Synonym palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
PubChem CID 445638
ChEBI CHEBI:28716
SMILES CCCCCC\C=C/CCCCCCCC(O)=O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 5 mg
IUPAC Name (9Z)-hexadec-9-enoic acid
Formula Weight 853.9
Physical Form Powder
Chemical Name or Material Paclitaxel, From the Pacific Yew Tree
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paclitaxel
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Paclitaxel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.