Learn More
MedChemExpress PACAP (1-38), human, ovine, rat
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Purity | 99.78% |
| CAS | 137061-48-4 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 4534.26 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 33.33 mg/mL (7.35 mM∣Need ultrasonic) |
| Synonym | Pituitary Adenylate Cyclase Activating Polypeptide 38 |