Learn More
Thermo Scientific™ PAC-dA Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsDescription
PAC-dA CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyadenosine (dA) with Phenoxyacetyl (PAC) base protection, for use in oligonucleotide synthesis. The UltraMILD monomers - phenoxyacetyl (PAC)-dA, acetyl (Ac)-dC, and iso-propyl-phenoxyacetyl (iPr-PAC)-dG - were developed to alleviate the problem of modifiers and labels not withstanding prolonged exposure to strongly alkaline deprotection conditions. Cleavage and deprotection can be achieved in 4 hours at room temperature with 0.05M potassium carbonate in methanol. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N6-phenoxyacetyl-2'-deoxyadenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C48H54N7O8P
• Base: dA
• Base Protection: PAC
• Formula weight: 887.96 g/mol
• CAS number: 110543-74-3
• HPLC Purity: ≥98.0%
• 31P NMR Purity: ≥98.0%
Specifications
Specifications
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | PAC-dA CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Deoxyadenosine (dA) |
| Reagent Features | Fast deprotection |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.