Learn More
PAC-1, MedChemExpress
Product Code. 16491887
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491887 50 mg 50mg
16465728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16491887 Supplier Medchem Express Supplier No. HY13523
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PAC-1 is a procaspase-3 activator that induces apoptosis in cancer cells with an EC50 of 2.08 μM.
PAC-1 is a procaspase-3 activator that induces apoptosis in cancer cells with an EC50 of 2.08 μM.
Specifications
Chemical Name or Material PAC-1
CAS 315183-21-2
Color White
Purity Grade Notes Research
Molecular Formula C23H28N4O2
Quantity 50 mg
Synonym Procaspase activating compound 1
Solubility Information DMSO : 50 mg/mL (127.39 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N/N=C/C1=CC=CC(CC=C)=C1O)CN2CCN(CC3=CC=CC=C3)CC2
Molecular Weight (g/mol) 392.49
Formula Weight 392.49
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death