missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70 S6 Kinase/S6K Antibody (5G9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00 €
Specifications
| Antigen | p70 S6 Kinase/S6K |
|---|---|
| Clone | 5G9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
p70 S6 Kinase/S6K Monoclonal specifically detects p70 S6 Kinase/S6K in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| p70 S6 Kinase/S6K | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| 6198 | |
| Purified recombinant fragment of human S6K expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 5G9 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Autophagy, Cancer, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
| EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A | |
| RPS6KB1 | |
| IgG1 | |
| Affinity Purified | |
| 59 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title